Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse Dynactin Subunit 1/DCTN1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6657SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6657SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6657SP Supplier R&D Systems Supplier No. MAB6657SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Dynactin Subunit 1/DCTN1 Monoclonal specifically detects Dynactin Subunit 1/DCTN1 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen Dynactin Subunit 1/DCTN1
Applications Western Blot
Classification Monoclonal
Clone 705007
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q14203
Gene Alias DAP-150, DCTN1, DP-150, Dynactin 1, dynactin 1 (p150, Glued (Drosophila) homolog), dynactin subunit 1, glued homolog, Drosophila), HMN7B, p135, p150 Glued (Drosophila) homolog, p150-glued
Gene Symbols DCTN1
Host Species Mouse
Immunogen E. coli-derived recombinant human Dynactin Subunit 1/DCTN1 Ala1145-Ser1278 Accession # Q14203
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1639
Test Specificity Detects human Dynactin Subunit 1/DCTN1 in direct ELISAs, and huamn and mouse DCTN1 in Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human DCTN2 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.