Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Dynactin Subunit 1/DCTN1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6657
This item is not returnable.
View return policy
Description
Dynactin Subunit 1/DCTN1 Monoclonal specifically detects Dynactin Subunit 1/DCTN1 in Human, Mouse samples. It is validated for Western Blot.Specifications
Dynactin Subunit 1/DCTN1 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
Q14203 | |
DCTN1 | |
E. coli-derived recombinant human Dynactin Subunit 1/DCTN1 Ala1145-Ser1278 Accession # Q14203 | |
100 μg | |
Primary | |
Detects human Dynactin Subunit 1/DCTN1 in direct ELISAs, and huamn and mouse DCTN1 in Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human DCTN2 is observed. | |
Human, Mouse | |
Purified |
Western Blot | |
705007 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
DAP-150, DCTN1, DP-150, Dynactin 1, dynactin 1 (p150, Glued (Drosophila) homolog), dynactin subunit 1, glued homolog, Drosophila), HMN7B, p135, p150 Glued (Drosophila) homolog, p150-glued | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1639 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction