Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse E-Cadherin Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. AF748 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF748 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF748 Supplier Bio-techne Sales Corp Supplier No. AF748
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

E-Cadherin Polyclonal specifically detects E-Cadherin in Human∣Mouse∣Pig∣Rat∣Transgenic Mouse∣Xenopus species samples. It is validated for Confocal Microscopy∣CyTOF∣Dual RNAScope ISH-IHC Compatible∣Flow Cytometry∣IF/IHC∣IFC∣Immunocytochemistry∣Immunofluorescence∣Immunohistochemistry∣Immunohistochemistry (Frozen)∣Immunohistochemistry (Paraffin)∣Neutralization∣Proximity Ligation Assay (PLA)∣Simple Western∣Western Blot.

Specifications

Antigen E-Cadherin
Applications Confocal Microscopy, CyTOF, Dual RNAScope ISH-IHC, Flow Cytometry, Immunofluorescence, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Neutralization, Proximity Ligation Assay, Simple Western, Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL, Simple Western 5 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, Immunocytochemistry 5-15 ug/mL, CyTOF-ready
Gene Accession No. P09803
Gene Alias Arc-1, CAD1, cadherin 1, E-cadherin (epithelial), cadherin 1, type 1, E-cadherin (epithelial), cadherin-1, calcium-dependent adhesion protein, epithelial, CAM 120/80, CD324, CD324 antigen, CDH1, CDHE, cell-CAM 120/80, Cell-CAM120/80, ECAD, E-cadherin, ECadherin, Epithelial cadherin, LCAM, L-CAM, UVOE-Cadherin, uvomorulin
Gene Symbols CDH1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse E-Cadherin Asp157-Val709 Accession # P09803
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 999
Test Specificity Detects mouse E-Cadherin in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Pig, Rat, Transgenic Mouse, Xenopus species
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.