Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse EBF-3 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF5166SP
This item is not returnable.
View return policy
Description
EBF-3 Polyclonal specifically detects EBF-3 in Human, Mouse samples. It is validated for Western Blot.Specifications
EBF-3 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
COE3, COE3OE-2, DKFZp667B0210, early B-cell factor 3O/E-2, EBF3, EBF-3, OE-2, Olf-1/EBF-like 2, transcription factor COE3 | |
Goat | |
Affinity Purified | |
RUO | |
253738 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 2 ug/mL | |
O08791-2 | |
EBF3 | |
E. coli-derived recombinant mouse EBF-3 Ile399-Ser504 Accession # O08791-2 | |
25 μg | |
Primary | |
Detects endogenous human and mouse EBF-3 in Western blots. | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction