Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse EBF-3 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF5166
This item is not returnable.
View return policy
Description
EBF-3 Polyclonal specifically detects EBF-3 in Human, Mouse samples. It is validated for Western Blot.Specifications
EBF-3 | |
Polyclonal | |
Unconjugated | |
O08791-2 | |
EBF3 | |
E. coli-derived recombinant mouse EBF-3 Ile399-Ser504 Accession # O08791-2 | |
100 μg | |
Primary | |
Detects endogenous human and mouse EBF-3 in Western blots. | |
Human, Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 2 ug/mL | |
COE3, COE3OE-2, DKFZp667B0210, early B-cell factor 3O/E-2, EBF3, EBF-3, OE-2, Olf-1/EBF-like 2, transcription factor COE3 | |
Goat | |
Affinity Purified | |
RUO | |
253738 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction