Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse EGLN1/PHD2 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$200.50 - $479.50
Specifications
Antigen | EGLN1/PHD2 |
---|---|
Clone | 2445D |
Dilution | Western Blot 1 μg/mL, Immunohistochemistry 3-25 μg/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB76801SP
![]() |
R&D Systems
MAB76801SP |
25 μg |
Each for $200.50
|
|
|||||
MAB76801100
![]() |
R&D Systems
MAB76801100 |
100 μg |
Each for $479.50
|
|
|||||
Description
EGLN1/PHD2 Monoclonal specifically detects EGLN1/PHD2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
EGLN1/PHD2 | |
Western Blot 1 μg/mL, Immunohistochemistry 3-25 μg/mL | |
Monoclonal | |
Supernatant | |
Human, Mouse | |
Q9QZT9 | |
54583 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
2445D | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
C1orf12, C1orf12HPH2, EC 1.14.11, EC 1.14.11.-, ECYT3, egl nine homolog 1, egl nine homolog 1 (C. elegans), egl nine-like protein 1, HIF prolyl hydroxylase 2, HIFPH2, HIF-PH2, HIFPH2 EGL nine (C.elegans) homolog 1, HIF-prolyl hydroxylase 2, HPH2, HPH-2, Hypoxia-inducible factor prolyl hydroxylase 2, PHD2, PHD2SM20, Prolyl hydroxylase domain-containing protein 2, SM20, SM-20, zinc finger MYND domain-containing protein 6, ZMYND6, ZMYND6DKFZp761F179 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human EGLN1/PHD2, Ala2-Phe426, Accession # Q9QZT9 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human EGLN1/PHD2 in direct ELISAs. Detects human and mouse EGLN1/PHD2 in Western blots. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title