Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse ELF3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB5787SP
This item is not returnable.
View return policy
Description
ELF3/ESE-1 Monoclonal specifically detects ELF3/ESE-1 in Human, Mouse samples. It is validated for Western Blot.Specifications
ELF3/ESE-1 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
P78545 | |
ELF3 | |
E. coli-derived recombinant human ELF3 Met1-Gly173 Accession # P78545 | |
25 μg | |
Primary | |
Detects human and mouse ELF3 in Western blots. | |
Human, Mouse | |
Purified |
Western Blot | |
662533 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
E74-like factor 3, E74-like factor 3 (ets domain transcription factor, epithelial-specific ), Epithelial-restricted with serine box, epithelial-specific), Epithelium-restricted Ets protein ESX, Epithelium-specific Ets transcription factor 1, EPR1, EPR-1, ERT, ESX, ets domain transcription factor, serine box (epithelial-specific), ETS-related transcription factor Elf-3, JEN, serine box | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1999 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction