Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse EMX2 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF6470SP
This item is not returnable.
View return policy
Description
EMX2 Polyclonal specifically detects EMX2 in Human, Mouse samples. It is validated for Western Blot.Specifications
EMX2 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
empty spiracles homeobox 2, Empty spiracles homolog 2, empty spiracles homolog 2 (Drosophila), Empty spiracles-like protein 2, homeobox protein EMX2 | |
Sheep | |
Affinity Purified | |
RUO | |
2018 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
Q04743 | |
EMX2 | |
E. coli-derived recombinant human EMX2 Asn49-Ser143 Accession # Q04743 | |
25 μg | |
Primary | |
Detects human EMX2 in Western blots. | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction