Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse EphA5 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $549.50
Specifications
Antigen | EphA5 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, CyTOF |
Classification | Polyclonal |
Description
EphA5 Polyclonal specifically detects EphA5 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
EphA5 | |
Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
Polyclonal | |
Goat | |
Human, Mouse | |
Bsk, Cek7, EC 2.7.10, EC 2.7.10.1, Ehk1, EHK-1, EHK1Hek7, EK7, EPH homology kinase 1, Eph homology kinase-1, EPH receptor A5, EphA5, EPH-like kinase 7, ephrin type-A receptor 5, hEK7, receptor protein-tyrosine kinase HEK7, Rek7, TYRO4HEK7CEK7, tyrosine-protein kinase receptor EHK-1 | |
EPHA5 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Flow Cytometry, CyTOF | |
Unconjugated | |
RUO | |
Q60629 | |
2044 | |
Mouse myeloma cell line NS0-derived recombinant mouse EphA5 Ser58-Pro412 Accession # Q60629 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse EphA5 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 15% cross-reactivity with recombinant mouse (rm) EphA3 is observed, 10% cross-reactivity with rmEphA7 is observed, 5% cross-reactivity with rmEphA2, rmEphA4, rmEphA8, rmEphB2, rmEphB3, and recombinant rat EphB1 is observed, and less than 1% cross-reactivity with rmEphA1, rmEphB4, and rmEphB6 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title