Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse EphB2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF467SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF467SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF467SP Supplier R&D Systems Supplier No. AF467SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 35 publications

EphB2 Polyclonal specifically detects EphB2 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready.

Specifications

Antigen EphB2
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Simple Western 20 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 1-15 ug/mL, Immunocytochemistry 5-15 ug/mL, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P54763
Gene Alias CAPB, Cek5, Drt, DRTEphB2, EC 2.7.10, EC 2.7.10.1, EK5, elk-related tyrosine kinase, EPH receptor B2, eph tyrosine kinase 3, EPH-like kinase 5, ephrin type-B receptor 2, EPHT3MGC87492, EPTH3, Erk, ERKHek5, HEK5, Nuk, PCBC, protein-tyrosine kinase HEK5, Qek2, Renal carcinoma antigen NY-REN-47, Sek3, Tyro5, Tyrosine-protein kinase receptor EPH-3, Tyrosine-protein kinase TYRO5
Gene Symbols EPHB2
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse EphB2 Val27-Lys548 Accession # P54763
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2048
Test Specificity Detects mouse and human EphB2 in direct ELISAs and Western blots. In Western blots, approximately 5% cross-reactivity with recombinant rat (rr) EphB1, recombinant mouse (rm) EphA8, rmEphA6, rmEphB6, rmEphA3, rmEphA4, rmEphA7, rrEphA5, recombinant human EphA1, rmEphA2 and rmEphB3 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.