Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse EphB2 PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB467P
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB467P 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB467P Supplier R&D Systems Supplier No. FAB467P
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

EphB2 Monoclonal specifically detects EphB2 in Human, Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen EphB2
Applications Flow Cytometry
Classification Monoclonal
Clone 512012
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. P54763
Gene Alias CAPB, Cek5, Drt, DRTEphB2, EC 2.7.10, EC 2.7.10.1, EK5, elk-related tyrosine kinase, EPH receptor B2, eph tyrosine kinase 3, EPH-like kinase 5, ephrin type-B receptor 2, EPHT3MGC87492, EPTH3, Erk, ERKHek5, HEK5, Nuk, PCBC, protein-tyrosine kinase HEK5, Qek2, Renal carcinoma antigen NY-REN-47, Sek3, Tyro5, Tyrosine-protein kinase receptor EPH-3, Tyrosine-protein kinase TYRO5
Gene Symbols EPHB2
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse EphB2 Val27-Lys548 (predicted) Accession # P54763
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2048
Test Specificity Detects human and mouse EphB2 in direct ELISAs.
Target Species Human, Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.