Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse ERK5/BMK1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2848
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2848 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF2848 Supplier R&D Systems Supplier No. AF2848
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 3 publications

ERK5/BMK1 Polyclonal specifically detects ERK5/BMK1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen ERK5/BMK1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunocytochemistry 10-25 ug/mL
Gene Accession No. Q13164
Gene Alias Big MAP kinase 1, BMK1, BMK-1, BMK1 kinase, BMK1EC 2.7.11.24, EC 2.7.11, ERK-5, ERK5MAPK 7, Extracellular signal-regulated kinase 5, extracellular-signal-regulated kinase 5, MAP kinase 7, MAPK7, mitogen-activated protein kinase 7, PRKM7ERK4
Gene Symbols MAPK7
Host Species Goat
Immunogen E. coli-derived recombinant human ERK5/BMK1 Met1-Glu432 Accession # Q13164
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5598
Test Specificity Detects human and mouse ERK5/BMK1 in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.