Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse EZH2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF4767
This item is not returnable.
View return policy
Description
EZH2/KMT6 Polyclonal specifically detects EZH2/KMT6 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry.Specifications
EZH2/KMT6 | |
Polyclonal | |
Unconjugated | |
Q15910 | |
EZH2 | |
E. coli-derived recombinant human EZH2 Gly512-Ile645 Accession # Q15910 | |
100 μg | |
Primary | |
Detects human and mouse EZH2 in direct ELISAs and Western blots. | |
Human, Mouse | |
IgG |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
EC 2.1.1, EC 2.1.1.43, enhancer of zeste (Drosophila) homolog 2, enhancer of zeste 2, Enhancer of zeste homolog 2, enhancer of zeste homolog 2 (Drosophila), ENX1, ENX-1KMT6EZH1KMT6A, histone-lysine N-methyltransferase EZH2, KMT6, Lysine N-methyltransferase 6, MGC9169 | |
Goat | |
Affinity Purified | |
RUO | |
2146 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction