Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse FABP2/I-FABP Antibody, R&D Systems™

Mouse Monoclonal Antibody
$208.00 - $1973.00
Specifications
Antigen | FABP2/I-FABP |
---|---|
Clone | 323730 |
Dilution | Western Blot 2 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB30781SP
![]() |
R&D Systems
MAB30781SP |
25 μg |
Each for $208.00
|
|
|||||
MAB30781100
![]() |
R&D Systems
MAB30781100 |
100 μg |
Each for $546.00
|
|
|||||
MAB30781500
![]() |
R&D Systems
MAB30781500 |
500 μg |
Each for $1,973.00
|
|
|||||
Description
FABP2/I-FABP Monoclonal specifically detects FABP2/I-FABP in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, ELISA Capture (Matched Antibody Pair).Specifications
FABP2/I-FABP | |
Western Blot 2 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FABPI, fatty acid binding protein 2, intestinal, Fatty acid-binding protein 2, fatty acid-binding protein, intestinal, IFABP, I-FABP, I-FABPMGC133132, Intestinal FABP, Intestinal-type fatty acid-binding protein | |
FABP2 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
323730 | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
Human, Mouse | |
P12104 | |
2169 | |
E. coli-derived recombinant human FABP2/I-FABP Ala2-Asp132 Accession # P12104 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human FABP2/I-FABP in ELISAs. Detects human and mouse FABP2/I-FABP in Western blots. In sandwich immunoassays, no cross-reactivity or interference with recombinant human FABP1, 3, 5, 6, 7, 8, 9, recombinant mouse FABP4, 9, or recombinant rat FABP2 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title