Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse FGF-8 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody has been used in 6 publications

$241.00 - $2020.00

Specifications

Antigen FGF-8
Clone 47109
Dilution Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Neutralization 0.25-0.75 ug/mL
Applications Western Blot, Immunohistochemistry, Neutralization
Classification Monoclonal
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB323SP
GREENER_CHOICE
View Documents
R&D Systems
MAB323SP
25 μg
Each for $241.00
Only null left
Add to Cart
 
MAB323100
GREENER_CHOICE
View Documents
R&D Systems
MAB323100
100 μg
Each for $549.50
Only null left
Add to Cart
 
MAB323500
GREENER_CHOICE
View Documents
R&D Systems
MAB323500
500 μg
Each for $2,020.00
Only null left
Add to Cart
 
Description

Description

FGF-8 Monoclonal specifically detects FGF-8 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.
Specifications

Specifications

FGF-8
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Neutralization 0.25-0.75 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
AIGF, AIGFKAL6, Androgen-induced growth factor, FGF8, FGF-8, fibroblast growth factor 8, fibroblast growth factor 8 (androgen-induced), HBGF-8, Heparin-binding growth factor 8, MGC149376
FGF8
IgG1
Protein A or G purified from ascites
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
47109
Western Blot, Immunohistochemistry, Neutralization
Unconjugated
Mouse
Human, Mouse
NP_001159834
2253
E. coli-derived recombinant mouse FGF-8b Gln23-Arg215 Accession # NP_001159834
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human and mouse FGF-8 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 2% cross-reactivity with recombinant human (rh) FGF-5, rhFGF-7, and rhFGF-9 is observed.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.