Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse FoxO3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6165
This item is not returnable.
View return policy
Description
FOXO3 Monoclonal specifically detects FOXO3 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry.Specifications
FOXO3 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunocytochemistry 5-25 ug/mL | |
O43524 | |
FOXO3 | |
E. coli-derived recombinant human FoxO3 Ala373-Gly673 Accession # O43524 | |
100 μg | |
Primary | |
Detects human and mouse FoxO3 in Western blots. | |
Human, Mouse | |
Purified |
Western Blot, Immunocytochemistry | |
648716 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
AF6q21, AF6q21 protein, DKFZp781A0677, FKHRL1, FKHRL1MGC31925, FKHRL1P2, forkhead box O3, forkhead box protein O3, forkhead homolog (rhabdomyosarcoma) like 1, Forkhead in rhabdomyosarcoma-like 1, forkhead, Drosophila, homolog of, in rhabdomyosarcoma-like 1, FOXO2, FOXO3A, FOXO3Aforkhead box O3A, MGC12739 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2309 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction