Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse FoxP2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5647
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5647 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5647 Supplier R&D Systems Supplier No. AF5647
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody has been used in 8 publications

FoxP2 Polyclonal specifically detects FoxP2 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen FoxP2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL
Gene Accession No. O15409
Gene Alias CAG repeat protein 44, CAGH44, CAGH44TNRC10forkhead box protein P2, DKFZp686H1726, forkhead box P2, forkhead/winged-helix transcription factor, SPCH1, TNRC10, trinucleotide repeat containing 10, Trinucleotide repeat-containing gene 10 protein
Gene Symbols FOXP2
Host Species Sheep
Immunogen E. coli-derived recombinant human FoxP2 isoform 1 Ala640-Glu715 Accession # O15409
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93986
Test Specificity Detects human and mouse FoxP2 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.