Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse FoxP3 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$233.50 - $538.00
Specifications
| Antigen | FoxP3 |
|---|---|
| Clone | 1054C |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FPK8214P025
![]() |
R&D Systems
FPK8214P025 |
25 Tests |
Each for $233.50
|
|
|||||
FPK8214P100
![]() |
R&D Systems
FPK8214P100 |
100 Tests |
Each for $538.00
|
|
|||||
Description
FoxP3 Monoclonal specifically detects FoxP3 in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
| FoxP3 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Rabbit | |
| Q9BZS1 | |
| 50943 | |
| E. coli-derived recombinant human FoxP3 Met1-Leu71 Accession # Q9BZS1 | |
| Primary | |
| Store the unopened product at 2 to 8 degreesC. Do not use past expiration date. Protect from light. |
| 1054C | |
| Flow Cytometry | |
| R-PE | |
| Human, Mouse | |
| AIID, AIIDMGC141961, DIETER, forkhead box P3, Forkhead Box Protein P3, FOXP3delta7, immune dysregulation, polyendocrinopathy, enteropathy, X-linked, Immunodeficiency, Polyendocrinopathy, Enteropathy, X-Linked, IPEX, JM2, MGC141961, MGC141963, PIDX, PIDXMGC141963, Scurfin, XPID, XPIDpolyendocrinopathy, enteropathy, X-linked | |
| FOXP3 | |
| IgG | |
| Affinity Purified | |
| Detects human and mouse FoxP3 in flow cytometry. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title