Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse FoxP3 PE-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems IC8214P100
This item is not returnable.
View return policy
Description
FoxP3 Monoclonal specifically detects FoxP3 in Human, Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
FoxP3 | |
Monoclonal | |
R-PE | |
Q9BZS1 | |
FOXP3 | |
E. coli-derived recombinant human FoxP3 Met1-Leu71 Accession # Q9BZS1 | |
100 Tests | |
50943 | |
Human, Mouse | |
Purified |
Flow Cytometry | |
1054C | |
Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
AIID, AIIDMGC141961, DIETER, forkhead box P3, Forkhead Box Protein P3, FOXP3delta7, immune dysregulation, polyendocrinopathy, enteropathy, X-linked, Immunodeficiency, Polyendocrinopathy, Enteropathy, X-Linked, IPEX, JM2, MGC141961, MGC141963, PIDX, PIDXMGC141963, Scurfin, XPID, XPIDpolyendocrinopathy, enteropathy, X-linked | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
Primary | |
Detects human and mouse FoxP3 in flow cytometry. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction