Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse Frizzled-2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1307SP
Change view
Click to view available options
Quantity:
25 μg
50 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1307SP 25 μg
MAB1307050 50 μg
MAB1307500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB1307SP Supplier R&D Systems Supplier No. MAB1307SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 1 publication

Frizzled-2 Monoclonal specifically detects Frizzled-2 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Frizzled-2
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 202102
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunoprecipitation
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9JIP6
Gene Alias frizzled (Drosophila) homolog 2, frizzled homolog 2 (Drosophila), frizzled-2, Frizzled2, fz-2, Fz2, FZD2, FzE2, hFz2
Gene Symbols FZD2
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Frizzled-2 Gln29-Leu168 Accession # Q9JIP6
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2535
Test Specificity Detects mouse Frizzled-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant mouse (rm) Frizzled-1, -3, -4, -6, -7, -8, or rhFrizzled-5.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.