Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse Frizzled-3 APC-conjugated Antibody, R&D Systems™

Catalog No. FAB1001A
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1001A 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB1001A Supplier R&D Systems Supplier No. FAB1001A

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Rat Monoclonal Antibody

Frizzled-3 Monoclonal specifically detects Frizzled-3 in Human, Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen Frizzled-3
Applications Flow Cytometry
Classification Monoclonal
Clone 169310
Conjugate APC
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q61086
Gene Alias frizzled (Drosophila) homolog 3, frizzled homolog 3 (Drosophila), frizzled-3, Frizzled3, Fz-3, FZD3, hFz3
Gene Symbols FZD3
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Frizzled-3 Met1-Val157 Accession # Q61086
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7976
Test Specificity Detects mouse Frizzled-3 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse Frizzled-1, -4, -7, or -8 is observed. By flow cytometry and immunohistochemistry, this antibody also detects mouse Frizzled-3 on differentiated embryoid bodies derived from the mouse embryonic cell line, D3. This antibody also detects Frizzled-3 on human HEK293 cells by immunocytochemistry and flow cytometry.
Target Species Human, Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.