Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse Frizzled-4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB194050
Change view
Click to view available options
Quantity:
100 μg
25 μg
50 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB194050 50 μg
MAB194SP 25 μg
MAB194100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB194050 Supplier R&D Systems Supplier No. MAB194050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 3 publications

Frizzled-4 Monoclonal specifically detects Frizzled-4 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready.

Specifications

Antigen Frizzled-4
Applications Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 145901
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q61088
Gene Alias CD344, CD344 antigen, EVR1, exudative vitreoretinopathy 1, FEVR, frizzled (Drosophila) homolog 4, frizzled homolog 4 (Drosophila), frizzled-4, Frizzled4, Fz-4, FZD4, FZD4S, FzE4, GPCR, hFz4, MGC34390, WNT receptor frizzled-4
Gene Symbols FZD4
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Frizzled-4 Phe37-Glu180 Accession # Q61088
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8322
Test Specificity Detects human and mouse Frizzled-4 in Western blots. In Western blots, no cross-reactivity with recombinant mouse (rm) Frizzled-1, -2, -3, -6, -7, -8, or recombinant human Frizzled-5 is observed. Detects Frizzled-4 by flow cytometry and immunohistochemistry on mouse differentiated embryoid bodies derived from the mouse embryonic cell line, D3.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.