Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse Frizzled-7 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 4 publications

Specifications

Antigen Frizzled-7
Clone 151143
Concentration LYOPH
Dilution Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready
Applications Flow Cytometry, Immunocytochemistry, CyTOF
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB1981050
View Documents
R&D Systems
MAB1981050
50 μg N/A N/A N/A
MAB1981100
View Documents
R&D Systems
MAB1981100
100 μg N/A N/A N/A
Description

Description

Frizzled-7 Monoclonal specifically detects Frizzled-7 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Specifications

Specifications

Frizzled-7
LYOPH
Flow Cytometry, Immunocytochemistry, CyTOF
Unconjugated
Rat
Human, Mouse
Q61090
8324
Mouse myeloma cell line NS0-derived recombinant mouse Frizzled-7 Gln33-Leu185 Accession # Q61090
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human and mouse Frizzled-7. In direct ELISAs, no cross-reactivity with recombinant mouse Frizzled-1, -3, -4 or -8 is observed.
151143
Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
frizzled (Drosophila) homolog 7, frizzled homolog 7 (Drosophila), Frizzled, drosophila, homolog of, 7, Frizzled7, fz-7, FZD7, FzE3frizzled-7, hFz7
FZD7
IgG2a
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.