Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse GDF-11/BMP-11 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF1958SP
This item is not returnable.
View return policy
Description
GDF-11/BMP-11 Polyclonal specifically detects GDF-11/BMP-11 in Human, Mouse samples. It is validated for Immunohistochemistry.Specifications
GDF-11/BMP-11 | |
Polyclonal | |
Immunohistochemistry 5-15 ug/mL | |
O95390 | |
GDF11 | |
E. coli-derived recombinant human GDF-11/BMP-11 Asn299-Ser407 Accession # O95390 | |
25 μg | |
Primary | |
Detects human and mouse GDF-11/BMP-11 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human/mouse/rat GDF-8/Myostatin and less than 1% cross-reactivity with recombinant human BMP-6 is observed. | |
Human, Mouse | |
IgG |
Immunohistochemistry | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
BMP11, BMP-11, BMP-11BMP11Bone morphogenetic protein 11, GDF11, GDF-11, growth differentiation factor 11, growth/differentiation factor 11 | |
Goat | |
Affinity Purified | |
RUO | |
10220 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction