Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse GDPD2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB7026SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7026SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB7026SP Supplier R&D Systems Supplier No. MAB7026SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

GDPD2 Monoclonal specifically detects GDPD2 in Human, Mouse samples. It is validated for Immunocytochemistry.

Specifications

Antigen GDPD2
Applications Immunocytochemistry
Classification Monoclonal
Clone 501016
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9HCC8
Gene Alias EC 3.1.4.43, FLJ20207, GDE3, GDE3Osteoblast differentiation promoting factor, Glycerophosphodiester phosphodiesterase 3, glycerophosphodiester phosphodiesterase domain containing 2, Glycerophosphodiester phosphodiesterase domain-containing protein 2, OBDPF, OBDPFglycerophosphoinositol inositolphosphodiesterase GDPD2
Gene Symbols GDPD2
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line transfected with human GDPD2 Accession # Q9HCC8
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54857
Test Specificity Detects human and mouse GDPD2.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.