Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Gephyrin/GPHN Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7519
This item is not returnable.
View return policy
Description
Gephyrin/GPHN Monoclonal specifically detects Gephyrin/GPHN in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Gephyrin/GPHN | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL | |
Q9NQX3 | |
GPHN | |
E. coli-derived recombinant human Gephyrin/GPHN Ala2-Ser188 Accession # Q9NQX3 | |
100 μg | |
Primary | |
Detects human Gephyrin/GPHN in direct ELISAs and human and mouse Gephyrin/GPHN Western blots. | |
Human, Mouse | |
Purified |
Western Blot, Immunohistochemistry | |
807423 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
GEPH, gephyrin, GPH, GPHN, GPHRYN, KIAA1385GPHGEPH | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
10243 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction