Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse GLI-3 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF3690
This item is not returnable.
View return policy
Description
GLI-3 Polyclonal specifically detects GLI-3 in Human, Mouse samples. It is validated for Western Blot.Specifications
GLI-3 | |
Polyclonal | |
Biotin | |
P10071 | |
GLI3 | |
E. coli-derived recombinant human GLI-3 Met1-Glu479 Accession # P10071 | |
50 μg | |
Primary | |
Detects human and mouse GLI-3 in Western blots. In Western blots, less than 1% cross-reactivity with recombinant human (rh) GLI-1 and rhGLI-2 is observed. | |
Human, Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
ACLS, ACLSPAPAPHS, ADD, Bph, GCPS, GLI family zinc finger 3, GLI3, GLI3 form of 190 kDa, GLI3 full length protein, GLI3-190, GLI3FL, GLI-Kruppel family member GLI3, glioma-associated oncogene family zinc finger 3, Greig cephalopolysyndactyly syndrome, oncogene GLI3, PAP-A, PAPA1, PAPB, PAPBDNA-binding protein, Pdn, PHS, PPDIV, Xt, zinc finger protein GLI3 | |
Goat | |
Affinity Purified | |
RUO | |
2737 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction