Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Glypican 5 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB2607T100UG
This item is not returnable.
View return policy
Description
Glypican 5 Monoclonal specifically detects Glypican 5 in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
| Glypican 5 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P78333 | |
| GPC5 | |
| Mouse myeloma cell line NS0-derived recombinant human Glypican 5 Glu25-Thr554 Accession # P78333 | |
| RUO | |
| 2262 | |
| Human, Mouse | |
| IgG2a |
| Flow Cytometry | |
| 297716 | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| bA93M14.1, glypican 5, glypican proteoglycan 5, glypican-5, GPC5 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human and mouse Glypican 5 in direct ELISAs and Western blots. In direct ELISAs and Western blots, 100% cross-reactivity with recombinant mouse Glypican 5 is observed and no cross-reactivity with recombinant human Glypican 2, 3 or 6 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction