Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse GSK-3 alpha/beta Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB25063
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB25063 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB25063 Supplier R&D Systems Supplier No. MAB25063
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

GSK-3 alpha/beta Monoclonal specifically detects GSK-3 alpha/beta in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen GSK-3 alpha/beta
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 833916
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Immunocytochemistry 3-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P49841
Gene Alias DKFZp686D0638, EC 2.7.11, EC 2.7.11.26, glycogen synthase kinase 3 alpha, glycogen synthase kinase-3 alpha, GSK3 alpha, GSK-3 alpha, GSK3 beta, GSK3A, GSK3B
Gene Symbols GSK3A
Host Species Mouse
Immunogen E. coli-derived recombinant human GSK-3 beta Asp200-Phe305 Accession # P49841
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2931
Test Specificity Detects human GSK-3 beta in direct ELISAs and human GSK-3 alpha and GSK-3 beta in Western blots. Detects human and mouse GSK-3 alpha/beta in immunocytochemistry.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.