Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse HAND2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3876
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3876 100 μg
AF3876SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF3876 Supplier R&D Systems Supplier No. AF3876
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 2 publications

HAND2 Polyclonal specifically detects HAND2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen HAND2
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. P61296
Gene Alias BHLHA26, bHLHa26MGC125303, dHAND, DHAND2, Ehand2, FLJ16260, heart and neural crest derivatives expressed 2, heart, autonomic nervous system and neural crestderivatives-expressed protein 2, Hed, MGC125304, Th2, Thing2
Gene Symbols HAND2
Host Species Goat
Immunogen E. coli-derived recombinant human HAND2 Met1-Gln217 Accession # P61296
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9464
Test Specificity Detects human and mouse HAND2 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human HAND1 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.