Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Haptoglobin Antibody, R&D Systems™

Goat Antibody
$200.50 - $549.50
Specifications
Antigen | Haptoglobin |
---|---|
Dilution | Western Blot 0.2 μg;/mL, Simple Western 1 μg;/mL, ELISA |
Applications | Western Blot, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Haptoglobin specifically detects Haptoglobin in Human, Mouse samples. It is validated for Western Blot, Simple Western, ELISA.Specifications
Haptoglobin | |
Western Blot, ELISA | |
Unconjugated | |
Goat | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
3240.0 | |
IgG | |
Antigen Affinity-purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Western Blot 0.2 μg;/mL, Simple Western 1 μg;/mL, ELISA | |
Polyclonal | |
Lyophilized | |
Human, Mouse | |
binding peptide, BP, haptoglobin, haptoglobin alpha(1S)-beta, haptoglobin alpha(2FS)-beta, haptoglobin, alpha polypeptide, haptoglobin, beta polypeptide, HP, HP-1, hp2-alpha, HP2ALPHA2, HPA1S, MGC111141, zonulin | |
Human Haptoglobin from human serum | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human and mouse Haptoglobin in Western blots. In sandwich immunoassays, human Haptoglobin is detected. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title