Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse Histone Deacetylase 8/HDAC8 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4359
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4359 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4359 Supplier R&D Systems Supplier No. AF4359
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Histone Deacetylase 8/HDAC8 Polyclonal specifically detects Histone Deacetylase 8/HDAC8 in Human, Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation (ChIP).

Specifications

Antigen Histone Deacetylase 8/HDAC8
Applications ChIP Assay
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Chromatin Immunoprecipitation (ChIP) 5 ug/5 x 10^6 cells
Gene Accession No. Q9BY41
Gene Alias EC 3.5.1.98, HD8, HDAC8, HDACL1, histone deacetylase 8, histone deacetylase-like 1, RPD3
Gene Symbols HDAC8
Host Species Sheep
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human HDAC8 isoform 1 Met1-Val377 (Leu31Pro) Accession # Q9BY41
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55869
Test Specificity Detects human HDAC8 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.