Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse HMGB1/HMG-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB16902100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB16902100 100 μg
MAB16902SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB16902100 Supplier R&D Systems Supplier No. MAB16902100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

HMGB1/HMG-1 Monoclonal specifically detects HMGB1/HMG-1 in Human, Mouse samples. It is validated for Immunocytochemistry.

Specifications

Antigen HMGB1/HMG-1
Applications Immunocytochemistry
Classification Monoclonal
Clone 1002147
Conjugate Unconjugated
Dilution Immunocytochemistry 8-25 μg/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P09429
Gene Alias Amphoterin, high mobility group box 1, High mobility group protein 1, high mobility group protein B1, high-mobility group (nonhistone chromosomal) protein 1, high-mobility group box 1, HMG1, HMG-1, HMG1DKFZp686A04236, HMG3, SBP-1, Sulfoglucuronyl carbohydrate binding protein
Host Species Mouse
Immunogen Synthetic peptide containing human HMGB1, Thr85-Gly166, Accession # P09429
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 3146
Test Specificity Detects human HMGB1 in direct ELISAs. Detects human and mouse HMGB1 in Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.