Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse HMGB3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5507
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5507 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB5507 Supplier R&D Systems Supplier No. MAB5507
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

HMGB3/HMG4 Monoclonal specifically detects HMGB3/HMG4 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen HMGB3/HMG4
Applications Western Blot
Classification Monoclonal
Clone 546737
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.05 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O15347
Gene Alias High mobility group protein 2a, High mobility group protein 4, high mobility group protein B3, high-mobility group (nonhistone chromosomal) protein 4, high-mobility group box 3, HMG-2a, HMG2A, HMG2AHMG-4, HMG4, HMG4MGC90319, non-histone chromosomal protein
Gene Symbols HMGB3
Host Species Mouse
Immunogen E. coli-derived recombinant human HMGB3 Met1-Val180 Accession # O15347
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3149
Test Specificity Detects human and mouse HMGB3 in Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.