Learn More
Human/Mouse HMGB3 Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | HMGB3/HMG4 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 ug/mL |
| Gene Accession No. | O15347 |
| Gene Alias | High mobility group protein 2a, High mobility group protein 4, high mobility group protein B3, high-mobility group (nonhistone chromosomal) protein 4, high-mobility group box 3, HMG-2a, HMG2A, HMG2AHMG-4, HMG4, HMG4MGC90319, non-histone chromosomal protein |
| Gene Symbols | HMGB3 |
| Host Species | Goat |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.