Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse HO-2/HMOX2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB3170
This item is not returnable.
View return policy
Description
HO-2/HMOX2 Monoclonal specifically detects HO-2/HMOX2 in Human, Mouse samples. It is validated for Western Blot.Specifications
HO-2/HMOX2 | |
Monoclonal | |
LYOPH | |
Western Blot 0.5 ug/mL | |
P30519 | |
HMOX2 | |
E. coli-derived recombinant human HO-2/HMOX2 Met1-Met316 Accession # P30519 | |
100 μg | |
Primary | |
Detects human and mouse HO-2/HMOX2 in Western blots. Endogenous rat HO-2 is detectable but has a higher background. In Western blots, no cross-reactivity with recombinant human HO-1/HMOX1 is observed. | |
Human, Mouse | |
Purified |
Western Blot | |
322913 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
heme oxygenase (decycling) 2, heme oxygenase 2, HMOX2, HO2, HO-2EC 1.14.99.3 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3163 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction