Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse IGF-I R/IGF1R Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 9 publications
Supplier: R&D Systems AF305SP
This item is not returnable.
View return policy
Description
IGF-I R/IGF1R Polyclonal specifically detects IGF-I R/IGF1R in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Neutralization, Immunocytochemistry.Specifications
IGF-I R | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD221, CD221 antigen, EC 2.7.10, EC 2.7.10.1, IGF1R, IGF-1R, IGF-I receptor, IGFIR, IGF-IR, IGFR, insulin-like growth factor 1 receptor, Insulin-like growth factor I receptor, JTK13, MGC142170, MGC142172, MGC18216, soluble IGF1R variant 1, soluble IGF1R variant 2 | |
Goat | |
Affinity Purified | |
RUO | |
3480 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry, Neutralization, Immunocytochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 0.5-1.5 ug/mL, Immunocytochemistry 1-15 ug/mL | |
P08069 | |
IGF1R | |
recombinant human IGF-I R extracellular domain Accession # P08069 | |
25 μg | |
Primary | |
Detects human and mouse IGF-I R in direct ELISAs and Western blots. In direct ELISAs and Western blots, 25-50% cross-reactivity with recombinant mouse IGF-I R is observed. In direct ELISAs, less than 1% cross-reactivity with recombinant human IGF-II R is observed. | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction