Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse IL-12/IL-35 p35 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$241.00 - $549.50
Specifications
Antigen | IL-12/IL-35p35 |
---|---|
Clone | 27537R |
Dilution | Western Blot 1μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB1570R-SP
![]() |
R&D Systems
MAB1570RSP |
25 μg |
Each for $241.00
|
|
|||||
MAB1570R-100
![]() |
R&D Systems
MAB1570R100 |
100 μg |
Each for $549.50
|
|
|||||
Description
IL-12/IL-35 p35 Monoclonal specifically detects IL-12/IL-35 p35 in Human, Mouse samples. It is validated for Western Blot.Specifications
IL-12/IL-35p35 | |
Western Blot 1μg/mL | |
Monoclonal | |
Supernatant | |
Human, Mouse | |
P29459 | |
IgG1 | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
27537R | |
Western Blot | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-12/IL-35 p35 Arg23-Ser219 Accession # P29459 | |
Primary | |
Reconstitute at 0.5mg/mL in sterile PBS. | |
Detects human IL-12/IL-35 p35 in direct ELISAs and Western blots. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title