Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse IL-12/IL-35 p35 APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Supplier: R&D Systems IC2191A
This item is not returnable.
View return policy
Description
IL-12/IL-35 p35 Monoclonal specifically detects IL-12/IL-35 p35 in Human, Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
IL-12/IL-35 p35 | |
Monoclonal | |
APC | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
IL12A | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-12/IL-35 p35 Arg23-Ser219 Accession # P29459 | |
100 Tests | |
Primary | |
Human, Mouse | |
Purified |
Flow Cytometry | |
27537 | |
Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
P29459 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Detects human IL-12/IL-35 p35 in direct ELISAs and Western blots. Detects the p35 subunit either as part of a p40/p35 heterodimer or as a free subunit after reduction of the heterodimer. This antibody does not recognize IL-12 p40 homodimers but shows strong cross-reactivity with the p35 subunits from porcine and mouse systems. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction