Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse IL-12 R beta 2 PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Supplier: R&D Systems FAB1959P100
This item is not returnable.
View return policy
Description
IL-12 R beta 2 Monoclonal specifically detects IL-12 R beta 2 in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
| IL-12 R beta 2 | |
| Monoclonal | |
| R-PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| IL-12 receptor beta 2, IL-12 receptor subunit beta-2, IL12R beta 2, IL-12R subunit beta-2, IL12RB2, IL-12RB2, IL-12R-beta-2, interleukin 12 receptor, beta 2, interleukin-12 receptor beta-2 chain, interleukin-12 receptor subunit beta-2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3595 | |
| Human, Mouse | |
| Purified |
| Flow Cytometry | |
| 305719 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Q99665 | |
| IL12RB2 | |
| Mouse myeloma cell line NS0-derived recombinant human IL-12 R beta 2 Cys28-Asn622 Accession # Q99665 | |
| 100 Tests | |
| Primary | |
| Detects human and mouse IL-12 R beta 2 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human IL-1 R beta 1 is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction