Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse IL-19 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
Supplier: R&D Systems MAB2915SP
This item is not returnable.
View return policy
Description
IL-19 Monoclonal specifically detects IL-19 in Human, Mouse samples. It is validated for Western Blot.Specifications
IL-19 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
Q8CJ70 | |
IL19 | |
E. coli-derived recombinant mouse IL-19 Leu25-Ala176 Accession # Q8CJ70 | |
25 μg | |
Primary | |
Detects human and mouse IL-19 in direct ELISAs and Western blots. | |
Human, Mouse | |
Purified |
Western Blot | |
350114 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
IL-10C, IL19, IL-19, interleukin 19, MDA1, melanoma differentiation associated protein-like protein, NG.1Melanoma differentiation-associated protein-like protein, ZMDA1interleukin-19 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
29949 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction