Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse IL-5 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems IC405T100UG
This item is not returnable.
View return policy
Description
IL-5 Monoclonal specifically detects IL-5 in Human, Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
IL-5 | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
BCDF mu, B-cell differentiation factor I, BCGFII, EDF, Eo-CSF, Eosinophil differentiation factor, IL5, IL-5T-cell replacing factor, interleukin 5 (colony-stimulating factor, eosinophil), interleukin-5, TRF, TRFB cell differentiation factor I | |
Rat | |
100 μg | |
Primary | |
Detects human and mouse IL-5 in ELISAs. In sandwich immunoassays with BAM6051 as the detection antibody, no cross-reactivity with recombinant human (rh) IL-3, rhGM-CSF, or recombinant mouse (rm) IL-5 is observed. In sandwich ELISAs with BAM705 as the detection antibody, no cross-reactivity with rhIL-5, rmIL-3, or rmGM-CSF is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Flow Cytometry | |
TRFK5 | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
IL5 | |
Mouse T cell clone-derived partially purified recombinant mouse IL-5 | |
RUO | |
3567 | |
Human, Mouse | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction