Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse IL-5 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Catalog No. IC405S100UG
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC405S100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC405S100UG Supplier R&D Systems Supplier No. IC405S100UG
This item is not returnable. View return policy

Rat Monoclonal Antibody

IL-5 Monoclonal specifically detects IL-5 in Human, Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen IL-5
Applications Flow Cytometry
Classification Monoclonal
Clone TRFK5
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 750
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Alias BCDF mu, B-cell differentiation factor I, BCGFII, EDF, Eo-CSF, Eosinophil differentiation factor, IL5, IL-5T-cell replacing factor, interleukin 5 (colony-stimulating factor, eosinophil), interleukin-5, TRF, TRFB cell differentiation factor I
Gene Symbols IL5
Host Species Rat
Immunogen Mouse T cell clone-derived partially purified recombinant mouse IL-5
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3567
Test Specificity Detects human and mouse IL-5 in ELISAs. In sandwich immunoassays with BAM6051 as the detection antibody, no cross-reactivity with recombinant human (rh) IL-3, rhGM-CSF, or recombinant mouse (rm) IL-5 is observed. In sandwich ELISAs with BAM705 as the detection antibody, no cross-reactivity with rhIL-5, rmIL-3, or rmGM-CSF is observed.
Target Species Human, Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.