Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse IL-5 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB405SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB405SP 25 μg
MAB405100 100 μg
MAB405500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB405SP Supplier R&D Systems Supplier No. MAB405SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 13 publications

IL-5 Monoclonal specifically detects IL-5 in Human, Mouse samples. It is validated for Neutralization, Intracellular Staining by Flow Cytometry, CyTOF-reported, ELISA Capture (Matched Antibody Pair), ELISA Capture (Matched Antibody Pair).

Specifications

Antigen IL-5
Applications Neutralization, Flow Cytometry, CyTOF, ELISA
Classification Monoclonal
Clone TRFK5
Conjugate Unconjugated
Dilution Neutralization 0.004-0.015 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-reported, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias BCDF mu, B-cell differentiation factor I, BCGFII, EDF, Eo-CSF, Eosinophil differentiation factor, IL5, IL-5T-cell replacing factor, interleukin 5 (colony-stimulating factor, eosinophil), interleukin-5, TRF, TRFB cell differentiation factor I
Gene Symbols IL5
Host Species Rat
Immunogen Mouse T cell clone-derived partially purified recombinant mouse IL-5
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3567
Test Specificity Detects human and mouse IL-5 in ELISAs. In sandwich immunoassays with BAM6051 as the detection antibody, no cross-reactivity with recombinant human (rh) IL-3, rhGM-CSF, or recombinant mouse (rm) IL-5 is observed. In sandwich ELISAs with BAM705 as the detection antibody, no cross-reactivity with rhIL-5, rmIL-3, or rmGM-CSF is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.