Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse ING1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB5758
This item is not returnable.
View return policy
Description
ING1 Monoclonal specifically detects ING1 in Human, Mouse samples. It is validated for Western Blot.Specifications
ING1 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
Q9UK53 | |
ING1 | |
E. coli-derived recombinant human ING1 Arg70-Ala184 Accession # Q9UK53 | |
100 μg | |
Primary | |
Detects human and mouse ING1 in Western blot. | |
Human, Mouse | |
Purified |
Western Blot | |
585915 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
growth inhibitor ING1, growth inhibitory protein ING1, inhibitor of growth family, member 1, inhibitor of growth protein 1, p24ING1c, p33, p33ING1, p33ING1b, p47, p47ING1a, tumor suppressor ING1 | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
3621 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction