Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Kirrel2/NEPH3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $514.50
Specifications
Antigen | Kirrel2/NEPH3 |
---|---|
Clone | 318311 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
Kirrel2/NEPH3 Monoclonal specifically detects Kirrel2/NEPH3 in Human, Mouse samples. It is validated for Western Blot.Specifications
Kirrel2/NEPH3 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse | |
Q6UWL6 | |
84063 | |
Mouse myeloma cell line NS0-derived recombinant human Kirrel2 Gly21-Arg503 Accession # Q6UWL6 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human and mouse Kirrel2/NEPH3 in direct ELISAs and Western blots. |
318311 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
DKFZp564A1164, Filtrin, FILTRINkin of IRRE-like protein 2, kin of IRRE like 2 (Drosophila), Kin of irregular chiasm-like protein 2, MGC15718, NEPH3, NEPH3Nephrin-like protein 3, nephrin-like 3, nephrin-like gene 1, NLG1, NLG1kin of irregular chiasm-like 2 | |
KIRREL2 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title