Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Layilin Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB2637G100UG
This item is not returnable.
View return policy
Description
Layilin Monoclonal antibody specifically detects Layilin in Human, Mouse samples. It is validated for Flow CytometrySpecifications
Layilin | |
Monoclonal | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
FLJ30977, FLJ31092, layilin, LAYN, LYAN | |
Mouse myeloma cell line NS0-derived recombinant human Layilin, Ala22-Glu220, Accession # NP_849156 | |
100 μg | |
Primary | |
Reconstitution default not known | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG1 |
Flow Cytometry | |
328024 | |
Flow Cytometry 0.25-1 ug/106 cells | |
NP_849156 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
143903 | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction