Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse LMO4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3889
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3889 100 μg
AF3889SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF3889 Supplier R&D Systems Supplier No. AF3889
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

LMO4 Polyclonal specifically detects LMO4 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen LMO4
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Gene Accession No. P61968
Gene Alias Breast tumor autoantigen, Crp3, Etohi4, LIM domain only 4, LIM domain only protein 4, LIM domain transcription factor LMO4, LMO-4
Gene Symbols LMO4
Host Species Sheep
Immunogen E. coli-derived recombinant human LMO4 Met1-Cys165 Accession # P61968
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8543
Test Specificity Detects human and mouse LMO4 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.