Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse LRRC4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4995
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4995 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB4995 Supplier R&D Systems Supplier No. MAB4995
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

LRRC4 Monoclonal specifically detects LRRC4 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen LRRC4
Applications Western Blot
Classification Monoclonal
Clone 701424
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9HBW1
Gene Alias BAG, Brain Tumor Associated Protein LRRC4, Brain Tumor-Associated Protein BAG, Leucine Rich Repeat Containing 4, Leucine-Rich Repeat-Containing 4, Leucine-Rich Repeat-Containing Protein 4, NAG14, Nasopharyngeal Carcinoma-Associated Gene 14 Protein, Netrin-G2 Ligand, NGL-2
Gene Symbols LRRC4
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human LRRC4 Ala39-Lys527 Accession # Q9HBW1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64101
Test Specificity Detects human and mouse LRRC4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) LRRC3, rhLRRC4B, or rhLRRC32 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.