Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse LRRN1/NLRR-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4990SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4990SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4990SP Supplier R&D Systems Supplier No. AF4990SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody has been used in 3 publications

LRRN1/NLRR-1 Polyclonal specifically detects LRRN1/NLRR-1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen LRRN1/NLRR-1
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. AAH34947
Gene Alias fibronectin type III, immunoglobulin and leucine rich repeat domains 3, FIGLER3, KIAA1497, leucine rich repeat neuronal 1, leucine-rich repeat neuronal protein 1, LRN, Neuronal leucine-rich repeat protein 1, NLRR1, NLRR-1
Gene Symbols LRRN1
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human LRRN1 Ser26-Ala631 Accession # AAH34947
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57633
Test Specificity Detects human LRRN1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant human NLRR-3 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.